missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Biohazard Identification Signs: Biohazard Infectious Agent Entry Requirements Contact Phone

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

$20.40 - $37.86

Specifications

Height (English) 14 in.
Height (Metric) 4.2 m
Width (English) 10 in.
Width (Metric) 3 m
Displayed Text BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
19140987
View Documents
Brady™
126646
Plastic
Each for $20.40
Only null left
 
19140993
View Documents
Brady™
126645
Aluminum
Each for $37.86
Only null left
 
Description

Description

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 14 in. H x 10 in. W
  • Legend: BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___

Specifications

Specifications

14 in.
10 in.
BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___
4.2 m
3 m
Biohazard Identification Sign
Videos
SDS
Documents

Documents