Learn More
DuPont™ Tychem™ 6000 Hooded Coverall
Catalog No. 19162822
Change view
Click to view available options
Size:
Small
Medium
Large
3X-Large
4X-Large
5X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19162822 Small
19162817 4X-Large
19162820 Large
19162816 3X-Large
19162821 Medium
19162819 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19162822 Supplier DuPont™ Supplier No. TF169TGYSM0006TV
Widely used by military personnel and first responders for chemical warfare situations
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Compliance
TAA Compliant
Specifications
Size Small
Gender Unisex
Material Tychem
Color Gray
Ankle Style Attached Socks
Closure Type Zipper
Seam Type Taped