Learn More
DuPont™ Tychem™ 6000 Hooded Coverall
Widely used by military personnel and first responders for chemical warfare situations
Supplier:  DuPont™ TF169TGY5X0006TV
 View more versions of this product
Catalog No. 19-162-819
Only null left
Explore available promotions
Description
Description
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Compliance
TAA Compliant
Specifications
Specifications
5X-Large
Tychem
Attached Socks
Taped
Unisex
Gray
Zipper
Product Suggestions
Product Suggestions