Learn More
DuPont™ Tychem™ 6000 Hooded Coverall
Widely used by military personnel and first responders for chemical warfare situations
Supplier: DuPont™ TF169TGY3X0006TV
Description
- Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Compliance
TAA Compliant
Specifications
| 3X-Large | |
| Tychem | |
| Attached Socks | |
| Taped |
| Unisex | |
| Gray | |
| Zipper |