missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mini Dry Bath, Accessory Blocks

Catalog No. 14955229
Change view
Click to view available options
Capacity (Metric):
0.2 mL
0.5 mL
1.5 mL
2 mL
15 mL
50 mL
No. of Places:
15
2
24
4
40
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) No. of Places
14955229 50 mL 2
14955225 2 mL 15
14955226 15 mL 4
14955224 1.5 mL 15
14955222 0.2 mL 40
14955223 0.5 mL 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14955229 Supplier Fisherbrand™ Supplier No. BSH100500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Compact, fits in the palm of your hand.

For use with Mini dry bath

For use with Mini dry bath

Order Info

For use with Mini dry bath

Specifications

Capacity (Metric) 50 mL
For Use With (Equipment) Mini Dry Bath
Material Aluminum
No. of Places 2
Type Block