Learn More
DuPont™ Tychem™ 6000 Hooded Coverall
Catalog No. p-4955706
Change view
Click to view available options
Size:
Small
3X-Large
5X-Large
6X-Large
7X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19162812 7X-Large
19162809 5X-Large
296356635 Small
296355397 3X-Large
19162811 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19162812 Supplier DuPont™ Supplier No. TF145TGY7X000600
Only null left
Widely used by military personnel and first responders for chemical warfare situations
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
Size 7X-Large
Gender Unisex
Material Tychem
Color Gray
Ankle Style Elastic
Closure Type Zipper
Seam Type Taped