missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Refrigerator/Freezer Ultra™ Thermometer

Catalog No. 361041622
Change view
Click to view available options
Probe Type:
1 Bottle and 1 Bullet Probe
1 Bottle Probe
1 Plastic Bottle Probe
1 x 5 mL Vaccine Bottle Probe
2 Plastic Bottle Probes
2 x 5 mL Vaccine Bottle Probe
One Bullet Probe
Two Bottle Probes
Two Bullet Probes
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Probe Type
361041622 1 Plastic Bottle Probe
14650103 1 Bottle Probe
14650104 Two Bottle Probes
14650105 One Bullet Probe
14650106 Two Bullet Probes
14650107 1 Bottle and 1 Bullet Probe
14650108 1 x 5 mL Vaccine Bottle Probe
14650109 2 x 5 mL Vaccine Bottle Probe
15079659 2 Plastic Bottle Probes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 361041622

Includes

Flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips

Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.

  • Designed to meet current CDC requirements for vaccine storage and monitoring
  • Triple display simultaneously shows min., max. and current temperatures for a specified probe
  • Smart-Alarm™ with visual and audible indicators signal continuously when an out-of-range condition is sensed
  • Alarm continues even after unit returns to non-alarm conditions and is programmable in 0.1° increments
  • °C/°F switchable
  • Enclosed temperature-buffered sensor to accurately monitor temperatures in one or two different areas in refrigerators, freezers, water baths and incubators
  • Solid-state probe sealed in a miniature bottle (1 x 2.5 in.) eliminates mercury contamination and rapid temperature changes when refrigerator door is opened
  • Miniature bottle is filled with nontoxic glycol, generally recognized as safe (GRAS) by FDA thereby eliminating concerns about contact with food or drinking water
  • Features high-impact, chemical-resistant ABS plastic case (2.75 x 4.25 x 0.75 in.)
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Range: -50° to +70°C (-58° to +158°F)
  • Accuracy: ±0.5°C between -30° to +50°C, otherwise ±1°C
  • Resolution: 0.1°
  • Sampling Rate: 30 seconds

Certifications

Individually serial-numbered Traceable™ Certificate is provided from ISO/IEC 17025:2005 calibration laboratory accredited by A2LA; indicates traceability to standards provided by NIST.

Specifications

Bottle Size 2.5 x 6.3 cm (1 x 2.5 in.)
Certifications/Compliance NIST Traceable
For Use With (Application) Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators
Includes Traceable™ Certificate, flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips
Length (Metric) 10.8 cm
Material Plastic
Probe Type 1 Plastic Bottle Probe
Resolution 0.1°
Weight (English) 4 oz.
Product Type Refrigerator/Freezer Ultra Thermometer
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
For Use With (Equipment) Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator
Accuracy ±0.5°C (-30°C to +50°C), ±1°C otherwise
Display Digital
Width (Metric) 6.99 cm
Show More Show Less