missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Refrigerator/Freezer Ultra™ Thermometer

Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.

Supplier:  Fisherbrand™ 9367811

 View more versions of this product

Catalog No. 14650109


Only null left

Includes: Flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips

Description

Description

Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.

  • Designed to meet current CDC requirements for vaccine storage and monitoring
  • Triple display simultaneously shows min., max. and current temperatures for a specified probe
  • Smart-Alarm™ with visual and audible indicators signal continuously when an out-of-range condition is sensed
  • Alarm continues even after unit returns to non-alarm conditions and is programmable in 0.1° increments
  • °C/°F switchable
  • Enclosed temperature-buffered sensor to accurately monitor temperatures in one or two different areas in refrigerators, freezers, water baths and incubators
  • Solid-state probe sealed in a miniature bottle (1 x 2.5 in.) eliminates mercury contamination and rapid temperature changes when refrigerator door is opened
  • Miniature bottle is filled with nontoxic glycol, generally recognized as safe (GRAS) by FDA thereby eliminating concerns about contact with food or drinking water
  • Features high-impact, chemical-resistant ABS plastic case (2.75 x 4.25 x 0.75 in.)
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Range: -50° to +70°C (-58° to +158°F)
  • Accuracy: ±0.5°C between -30° to +50°C, otherwise ±1°C
  • Resolution: 0.1°
  • Sampling Rate: 30 seconds

Certifications

Individually serial-numbered Traceable™ Certificate is provided from ISO/IEC 17025:2005 calibration laboratory accredited by A2LA; indicates traceability to standards provided by NIST.

Specifications

Chemical Identifiers

2.5 x 6.3 cm (1 x 2.5 in.)
0.1°
Refrigerator/Freezer Ultra Thermometer
70°C
-50°C
-50°C to +70°C
Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator
Digital
Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators
4 oz.
158°F
-58°F
-58°F to +158°F
Yes
±0.5°C (-30°C to +50°C), ±1°C otherwise
6.99 cm

Specifications

Nontoxic Glycol
NIST Traceable
Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators
7 cm
2 x 5 mL Vaccine Bottle Probe
4 oz.
Refrigerator/Freezer Ultra Thermometer
158°F
-58°F
-58°F to +158°F
Yes
±0.5°C (-30°C to +50°C), ±1°C otherwise
6.99 cm
2.5 x 6.3 cm (1 x 2.5 in.)
7 x 10.8 x 2 cm (2.75 x 4.25 x 0.75 in.)
Traceable™ Certificate, flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips
ABS Plastic
0.1°
113.4 g
3 in.
70°C
-50°C
-50°C to +70°C
Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator
Digital
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions