missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Refrigerator/Freezer Ultra™ Thermometer
Description
Includes
Flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips
Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.
- Designed to meet current CDC requirements for vaccine storage and monitoring
- Triple display simultaneously shows min., max. and current temperatures for a specified probe
- Smart-Alarm™ with visual and audible indicators signal continuously when an out-of-range condition is sensed
- Alarm continues even after unit returns to non-alarm conditions and is programmable in 0.1° increments
- °C/°F switchable
- Enclosed temperature-buffered sensor to accurately monitor temperatures in one or two different areas in refrigerators, freezers, water baths and incubators
- Solid-state probe sealed in a miniature bottle (1 x 2.5 in.) eliminates mercury contamination and rapid temperature changes when refrigerator door is opened
- Miniature bottle is filled with nontoxic glycol, generally recognized as safe (GRAS) by FDA thereby eliminating concerns about contact with food or drinking water
- Features high-impact, chemical-resistant ABS plastic case (2.75 x 4.25 x 0.75 in.)
- Ten-foot micro-cable permits refrigerator doors to close on it
- Range: -50° to +70°C (-58° to +158°F)
- Accuracy: ±0.5°C between -30° to +50°C, otherwise ±1°C
- Resolution: 0.1°
- Sampling Rate: 30 seconds
Certifications
Individually serial-numbered Traceable™ Certificate is provided from ISO/IEC 17025:2005 calibration laboratory accredited by A2LA; indicates traceability to standards provided by NIST.
Specifications
Specifications
| Bottle Fill | Nontoxic Glycol |
| Bottle Size | 2.5 x 6.3 cm (1 x 2.5 in.) |
| Certifications/Compliance | NIST Traceable |
| Dimensions (L x W x H) | 7 x 10.8 x 2 cm (2.75 x 4.25 x 0.75 in.) |
| For Use With (Application) | Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators |
| Includes | Traceable™ Certificate, flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips |
| Length (Metric) | 7 cm |
| Material | ABS Plastic |
| Probe Type | One Bullet Probe |
| Resolution | 0.1° |
| Show More |