missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Non-Combustible Outdoor Safety Lockers
Designed for storage of flammable and combustible liquids contained in chemical drums varying in capacities
Supplier: Justrite™ 911120
Description
- Made of heavy gauge steel (backed by 15 year structural warranty)
- Leakproof all welded sump secondary containment
- Removable, galvanized steel grating
- Three-point keyed locking hardware
- Natural draft ventilation
- Multiple static grounding connections
- Anchoring plates for maximum hold down protection
- Chemical and UV resistant coating, inside and out
- Forklift channels for easy relocation
- Available with and without Explosion Relief Panels
- NFPA 704 diamond decal provides Right to Know information
- FM-approved - including sump capacity and wind/snow load requirements
Model Specific Features:
- With Explosion relief panels designed to release at 20 psf (911021, 911041, 911061, 911091, 911121, and 911161)
- Without Explosion relief panels (911020, 911040, 911060, 911090, 911120 and 911160)
Certifications
FM approved
Compliance
Meets EPA, NFPA 30 and NFPA 1 requirements
Chemical Identifiers
| FM approved, EPA, NFPA 30, NFPA 1 compliant | |
| Bone | |
| Non-combustible Outdoor Safety Locker | |
| Hazardous/chemical storage |
| Heavy gauge steel | |
| Steel | |
| Three-point lock | |
| 1 |
Specifications
| Non-combustible Outdoor Safety Locker | |
| 198.1 cm | |
| 213.4 cm | |
| 2591 mm | |
| 1 | |
| 12 x 55 gal. drums | |
| Heavy gauge steel | |
| 102 in. | |
| 84 in. | |
| FM approved, EPA, NFPA 30, NFPA 1 compliant | |
| Steel | |
| 78 in. |
| 102 x 78 x 84 in. | |
| 246.4 cm | |
| 2921 mm | |
| 213.4 cm | |
| No | |
| 97 in. | |
| 84 in. | |
| Hazardous/chemical storage | |
| 115 in. | |
| Bone | |
| Three-point lock |