missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Miller™ AirCore™ Steel Harnesses
Description
- Lightweight, unique breathable, open-core padding technology provides optimal airflow and reduces heat and moisture build-up up
- Contoured ergonomic padding design minimizes contact around a worker's neck, lower back and inner thighs to reduce chafing and irritation
- Patented DuraFlex stretchable webbing provides greater flexibility, increased comfort
- Cam buckles provide easy one-hand adjustment
- Holds webbing firmly in place with no readjusting required
- Stand-up back D-ring easily adjusts and attaches
- Quick-connect buckles for easy donning
- Unique and vibrant patterned webbing for easy identification
- Available with quick connect (QC) or tongue buckle, side-D rings, back-D rings
- Recommended Industries/Use: Oil and Gas
- Hazards: Confined Space, fall from height
Certifications
OSHA, ANSI
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI A10.32 and ANSI Z359.1-2007; CSA Z259.10-12 requirements.
Specifications
Specifications
| Buckle Style (Leg) | Quick-Connect |
| Includes | Quick-Connect chest and leg |
| Capacity (Metric) | 181.5 kg |
| D-Ring Location | None |
| Buckle Style (Chest) | Quick-Connect |
| Capacity (English) | 400 lb. |