Learn More
Celltreat Scientific Products 48 Well Non-treated Plate, Sterile
Supplier:  Celltreat Scientific Products 229548
Encompass
CELLTREAT Scientific Products 48 Well Non-Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with working volume of 0.3-1.0mL. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 100/case.
Catalog No. 50202147
Only null left