missing translation for 'onlineSavingsMsg'
Learn More

Laxco™ 46° Brix Calibration Fluid for Handheld Analog Refractometers
GREENER_CHOICE

Catalog No. RPCAL46P
Change view
Click to view available options
Quantity:
Case of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RPCAL46P Case of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RPCAL46P Supplier Laxco™ Supplier No. RPCAL46P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

5ml droplet bottle of brix 46° solution

Used to Calibrate Handheld Analog Brix Refractometers

  • 5mL droplet bottle of 46° Brix Calibration solution for calibration of Handheld Analog Refractometer models for brix models range from 28° to 62°
  • Quantity: 3

Specifications

Type Calibration Fluid
Scale Range 46 Brix
For Use With (Equipment) Refractometer
Description 5mL bottle
Accuracy ±0.1°
Quantity Case of 5
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase