Learn More
V&P Scientific Inc MULTI-BLOT REPLICATOR, 48 or 96 Wells, Standard Plate, Flat Tip, Delivers ~3,000nL, 3.15mm Pin Diameter
Catalog No. NC0540875
Encompass
MULTI-BLOT REPLICATOR, 48 or 96 Wells, Standard Well Plate, Stainless Steel, Flat Tip, Delivers ~3,000nL Hanging Drop, 3.15mm Pin Diameter, 22mm Total Length, 9mm Spacing
Catalog No. NC0540875 Supplier V&P Scientific Inc Supplier No. VP 407AH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MULTI-BLOT REPLICATOR, 48 or 96 Wells, Standard Well Plate, Stainless Steel, Flat Tip, Delivers ~3,000nL Hanging Drop, 3.15mm Pin Diameter, 22mm Total Length, 9mm Spacing