4000ml Reaction Vessel, Cylindrical, 150mm Schott O-Ring Flange, 160mm O.D. x 355mm Height. Cylindrical reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T, for o-ring see CG-147.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4000ml Reaction Vessel, Cylindrical, 150mm Schott O-Ring Flange, 160mm O.D. x 355mm Height. Cylindrical reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T, for o-ring see CG-147.