Learn More
Chemglass Inc 4000mL Glas-col Heating Mantle, Series O, Cylindrical, 6 5/16in ID x 11 1/4in Depth, Bottom Drain Opening, 1000 Watts
Catalog No. CG1000918
Encompass
4000mL Glas-col Heating Mantle, Series O, Cylindrical, 6 5/16in ID x 11 1/4in Depth, Bottom Drain Opening, 1000 Watts. This Glas-Col Series O mantle is similar to CG-10007 except mantle is slit lengthwise to a 1 hole in the bottom center with lacing closure to accommodate a cylindrical reaction vessel, CG-1924, with a bottom drain. Mantle is glass fabric, 115 volts, and is supplied with a separable 4 foot, 2 wire extension cord with locking connector for each circuit. The 10,000mL size has two circuits at the listed wattage.
Catalog No. CG1000918 Supplier Chemglass Inc Supplier No. CG1000918
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4000mL Glas-col Heating Mantle, Series O, Cylindrical, 6 5/16in ID x 11 1/4in Depth, Bottom Drain Opening, 1000 Watts. This Glas-Col Series O mantle is similar to CG-10007 except mantle is slit lengthwise to a 1 hole in the bottom center with lacing closure to accommodate a cylindrical reaction vessel, CG-1924, with a bottom drain. Mantle is glass fabric, 115 volts, and is supplied with a separable 4 foot, 2 wire extension cord with locking connector for each circuit. The 10,000mL size has two circuits at the listed wattage.