missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ 40 Cal Side Fan Assembly For Revolite Hood

Catalog No. 17001688
Change view
Click to view available options
Style:
Left Side
Right Side
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
17001688 Left Side
17001690 Right Side
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17001688 Supplier National Safety Apparel™ Supplier No. FANNPLS

Specifications

Size Medium
Material Plastic
Color Black
Seam Type Stitched
For Use With (Equipment) Arc Flash Hoods
Includes Fan
Style Left Side