Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite Crossvent Hood with PureView Faceshield and Headlamp
Catalog No. 17001730
Encompass
Change view
Click to view available options
Color: Olive green
Catalog No.
17001730 Color: Olive green
1 options
Catalog No. 17001730 Supplier National Safety Apparel™ Supplier No. H65NPQHFANPVL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Crossvent hood with purview faceshield and headlamp
  • Arc Rating: 40 cal/cm²
  • CAT 4
  • Fabric/Material: 11.2 oz. RevoLite™
  • 50% lighter than our standard 40 cal/cm² Arc Flash PPE
  • Inherently flame resistant
  • Anti-fog faceshield
  • Fans are attached with hook & loop
  • Hood provides a gentle and quiet breeze that keeps you cool and decreases lens fogging
  • Hard hat with slotted adapter included
  • FR fabric will not melt, drip, or ignite
  • Headlamp provides additional visibility in low light settings
  • Made in the USA
Specifications
Size Medium
Material RevoLite™
Color Olive Green
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269