Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite™ Arc Flash Coat
Catalog No. 17001220
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001220 5X-Large
17001213 Small
17001214 Medium
17001215 Large
17001216 X-Large
17001217 2X-Large
17001218 3X-Large
17001219 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17001220 Supplier National Safety Apparel™ Supplier No. C04NPQHLTD5X32
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Arc flash coats are equipped with hook and loop front closure with drawstring waist.
  • 50% lighter than standard 40 cal/cm2 arc flash kits
  • Stand up collar for extra coverage, can be turned down
  • Nomex knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • FR fabric will not melt, drip, or ignite
  • Available in a convenient kit
Specifications
Closure Type FR Hook and Loop with Buckle
Color Olive Green
Disposable Reusable
Gender Unisex
Material RevoLite™
Neck Style Collared
Size 5X-Large
Resistance Flame
Seam Type Stitched
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Show More Show Less