missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ 40 Cal ArcGuard™ Compliance Arc Flash Leggings

Catalog No. p-7220117
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170012050 Large
170012051 X-Large
170012052 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 170012050 Supplier National Safety Apparel™ Supplier No. L05UQUQ40LG
Only null left

Compliance leggings with arc rating of 40 cal/cm2

  • 9 oz. Double Layer UltraSoft™
  • Adjustable FR hook and loop closure
  • For use with 40 cal FR long coat

Specifications

Size Large
Gender Unisex
Material Cotton, Nylon
Color Navy
Ankle Style Open
Seam Type Stitched
Disposable Reusable
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Sterility Non-sterile