missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 40 cal Arc Flash Kit (10)

Catalog No. p-7220229
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011624 Small
170011629 Medium
170011634 Large
170011639 X-Large
170011644 2X-Large
170011649 3X-Large
170011654 4X-Large
170011659 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011624 Supplier National Safety Apparel™ Supplier No. KIT4SC40SM10
Only null left

Kit Includes: 40 cal Compliance short coat & bib overall, Arc flash hood with PureView™ faceshield and hard hat, safety glasses, hood bag and gear bag

  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • FR cuffs improve fit with electrical gloves
  • PureView™ faceshield technology features a greater range of visibility
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots

Specifications

Product Type Arc Flash Kit
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Material Cotton, Nylon
Arc Flash Rating 40 cal/cm²
Closure Type FR Hook and Loop
Seam Type Stitched
Suspender Adjustable
Ankle Style Open
Color Navy
Size Small
Show More Show Less