Learn More
Celltreat Scientific Products 4 Well Tissue Culture Plate, Sterile
Supplier:  Celltreat Scientific Products 229103
Encompass
CELLTREAT Scientific Products 4 Well Tissue Culture Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. 4 well plate is a smaller footprint option which offers well size similar to that of a 24 well plate, but has expanded well spacing to reduce cross contamination. Has a growth area of 1.96cm. RNase/DNase Free and non pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 50/case.
Catalog No. 501149116
Only null left