Learn More
Thermo Scientific™ 4-Way Flipper™ Racks
GREENER_CHOICE
Catalog No. 2140227
Change view
Click to view available options
Color:
Fluorescent Green
Fluorescent Orange
Fluorescent Pink
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
2140227 Fluorescent Orange
2140228 Fluorescent Pink
2140226 Fluorescent Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2140227 Supplier Thermo Scientific™ Supplier No. 8860
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™4-Way Flipper™ Racks.
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™ 4-Way Flipper™ Racks.
Specifications
Description 4-Way Flipper Rack
Color Fluorescent Orange
Holds 0.5 mL, 15 mL, Tubes (50 mL)
Positions 32 x 0.5 mL, 32 x 1.5 mL, 12 x 15 mL or 4 x 50 mL tubes, each on a different side
Shape Rectangular
Product Type 4-way Flipper Rack