missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-2702173
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC359810010 1 g Glass bottle
AC359810050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC359810010 Supplier Thermo Scientific Chemicals Supplier No. 359810010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2156-04-9
Molecular Formula C8H9BO2
Molecular Weight (g/mol) 147.97
MDL Number MFCD00239441
InChI Key QWMJEUJXWVZSAG-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 190°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 16, IV, 1676
Synonym 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,dmso,dmf,thf,diethyl ether
IUPAC Name (4-ethenylphenyl)boronic acid
PubChem CID 2734393
Formula Weight 147.97
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Vinylbenzeneboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only