missing translation for 'onlineSavingsMsg'
Learn More

4-Vinyl-1-cyclohexene, 97%, stabilized

Catalog No. p-211376
Change view
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140880050 5 mL Glass bottle
AC140885000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC140880050 Supplier Thermo Scientific Chemicals Supplier No. 140880050
Only null left

CAS: 100-40-3 | C8H12 | 108.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-40-3
Molecular Formula C8H12
Molecular Weight (g/mol) 108.18
MDL Number MFCD00001576
InChI Key BBDKZWKEPDTENS-UHFFFAOYSA-N
Synonym 4-vinylcyclohexene, 4-vinyl-1-cyclohexene, cyclohexene, 4-ethenyl, butadiene dimer, 4-vinylcyclohex-1-ene, 4-vinylcyclohexene-1, cyclohexene, 4-vinyl, cyclohexenylethylene, 4-ethenyl-1-cyclohexene, 1-vinylcyclohexene-3
PubChem CID 7499
ChEBI CHEBI:82377
IUPAC Name 4-ethenylcyclohexene
SMILES C=CC1CCC=CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -101.0°C
Color Beige-Brown to White
Density 0.8320g/mL
Boiling Point 126.0°C to 127.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4625 to 1.4645
Linear Formula C6H9CH=CH2
Quantity 5 mL
Beilstein 05, I, 63
Specific Gravity 0.832
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in benzene and ether
Stabilizer 25 to 200ppm BHT
Viscosity 0.7 mPa.s (20°C)
Formula Weight 108.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Vinyl-1-cyclohexene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENT
EINECSNumber 202-848-9
RTECSNumber GW6650000
TSCA TSCA

RUO – Research Use Only