missing translation for 'onlineSavingsMsg'
Learn More

4-(Trimethylsilyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T26641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26641G 1 g
T26645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26641G Supplier TCI America Supplier No. T26641G

Chemical Identifiers

CAS 17865-11-1
Molecular Formula C9H15BO2Si
Molecular Weight (g/mol) 194.11
MDL Number MFCD00093348
InChI Key NRPZMSUGPMYBCQ-UHFFFAOYSA-N
Synonym 4-(Trimethylsilyl)benzeneboronic Acid
PubChem CID 1710
IUPAC Name [4-(trimethylsilyl)phenyl]boronic acid
SMILES C[Si](C)(C)C1=CC=C(C=C1)B(O)O

Specifications

Color White
Quantity 1 g
Formula Weight 194.11
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trimethylsilyl)phenylboronic Acid (contains varying amounts of Anhydride)