missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trimethylsilyl)diphenylacetylene 97.0+%, TCI America™
Supplier: TCI America T25965G
Specifications
| 4-(Trimethylsilyl)diphenylacetylene | |
| 39°C | |
| C17H18Si | |
| 5 g | |
| UZRJWRFLYPENFH-UHFFFAOYSA-N | |
| trimethyl-[4-(2-phenylethynyl)phenyl]silane | |
| 21093331 | |
| ≥97.0% (GC) |
| 136459-72-8 | |
| White-Yellow | |
| MFCD09953470 | |
| 4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene | |
| C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2 | |
| 250.416 | |
| 250.42 | |
| Crystalline Powder |
Chemical Identifiers
| 136459-72-8 | |
| 250.416 | |
| UZRJWRFLYPENFH-UHFFFAOYSA-N | |
| 21093331 | |
| C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2 |
| C17H18Si | |
| MFCD09953470 | |
| 4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene | |
| trimethyl-[4-(2-phenylethynyl)phenyl]silane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator