missing translation for 'onlineSavingsMsg'
Learn More

4-(Trimethylsilyl)diphenylacetylene, 99%

CAS: 136459-72-8 | C17H18Si | 250.416 g/mol

Supplier:  Thermo Scientific Chemicals H2630006

 View more versions of this product

Catalog No. AAH2630006


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Trimethylsilyl)diphenylacetylene
136459-72-8
C17H18Si
4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene
C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2
250.416
250.42
37°C to 39°C
5 g
MFCD09953470
UZRJWRFLYPENFH-UHFFFAOYSA-N
trimethyl-[4-(2-phenylethynyl)phenyl]silane
21093331
99%

Chemical Identifiers

136459-72-8
250.416
UZRJWRFLYPENFH-UHFFFAOYSA-N
21093331
C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2
C17H18Si
MFCD09953470
4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene
trimethyl-[4-(2-phenylethynyl)phenyl]silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only