missing translation for 'onlineSavingsMsg'
Learn More

4-(Trimethylsilyl)diphenylacetylene, 99%

Catalog No. AAH2630003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2630003 1 g
AAH2630006 5 g
AAH2630014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2630003 Supplier Thermo Scientific Chemicals Supplier No. H2630003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 136459-72-8 | C17H18Si | 250.416 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136459-72-8
Molecular Formula C17H18Si
Molecular Weight (g/mol) 250.416
MDL Number MFCD09953470
InChI Key UZRJWRFLYPENFH-UHFFFAOYSA-N
Synonym 4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene
PubChem CID 21093331
IUPAC Name trimethyl-[4-(2-phenylethynyl)phenyl]silane
SMILES C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2

Specifications

Melting Point 37°C to 39°C
Quantity 1 g
Formula Weight 250.42
Percent Purity 99%
Chemical Name or Material 4-(Trimethylsilyl)diphenylacetylene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only