missing translation for 'onlineSavingsMsg'
Learn More

4-Trimethylsilyl-3-butyn-1-ol, 98%

Catalog No. p-7035657
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
AAH5337606 5 g
AAH5337614 25 g
2 options

Catalog No. AAH5337606

Supplier: Thermo Scientific Chemicals H5337606

Only null left

CAS: 2117-12-6 | C7H14OSi | 142.27 g/mol

4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2117-12-6
MDL Number MFCD00190217

Specifications

Boiling Point 97°C to 98°C (24mmHg)
Quantity 5 g
Molecular Formula C7H14OSi
Linear Formula HOCH2CH2C≡CSi(CH3)3
UN Number UN1993
Flash Point 60°C (140°F)
Synonym 4-trimethylsilyl-3-butyn-1-ol, 4-trimethylsilyl but-3-yn-1-ol, 3-butyn-1-ol, 4-trimethylsilyl, 4-trimethylsilyl-3-butyne-1-ol, 1-trimethylsilyl-1-butyn-4-ol, 3-butyn-1-ol,4-trimethylsilyl
InChI Key VSTMVAFCUIGHQI-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CCCO
IUPAC Name 4-trimethylsilylbut-3-yn-1-ol
Molecular Weight (g/mol) 142.27
PubChem CID 2760827
Formula Weight 142.27
Percent Purity 98%
Density 0.854
Chemical Name or Material 4-Trimethylsilyl-3-butyn-1-ol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only