missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)pyridine, 97%

Catalog No. AAB2295106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2295106 5 g
AAB2295103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2295106 Supplier Thermo Scientific Chemicals Supplier No. B2295106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3796-24-5 | C6H4F3N | 147.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3796-24-5
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00153289
InChI Key IIYVNMXPYWIJBL-UHFFFAOYSA-N
Synonym 4-trifluoromethyl pyridine, pyridine, 4-trifluoromethyl, 4-trifluoromethyl-pyridine, pubchem8028, p-trifluoromethylpyridine, acmc-1ctve
PubChem CID 138055
IUPAC Name 4-(trifluoromethyl)pyridine
SMILES C1=CN=CC=C1C(F)(F)F

Specifications

Density 1.27
Boiling Point 107°C to 110°C
Flash Point 20°C (68°F)
Refractive Index 1.4147
Quantity 5 g
UN Number UN1993
Beilstein 1364589
Formula Weight 147.1
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)pyridine
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only