missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)piperidine hydrochloride, 97%

Catalog No. p-7047556
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19441MD 250 mg
AAL1944103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19441MD Supplier Thermo Scientific Chemicals Supplier No. L19441MD

CAS: 155849-49-3 | C6H11ClF3N | 189.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 155849-49-3
Molecular Formula C6H11ClF3N
Molecular Weight (g/mol) 189.606
MDL Number MFCD04971993
InChI Key KMUKXPQVPQUCDB-UHFFFAOYSA-N
Synonym 4-trifluoromethyl piperidine hydrochloride, 4-trifluoromethyl piperidine hcl, 4-trifluoromethyl piperidinehydrochloride, 4-trifluoromethylpiperidine hydrochloride, 4-trifluoromethyl-piperidine hydrochloride, piperidine, 4-trifluoromethyl-, hydrochloride, 4-trifluoromethyl piperidine, chloride, pubchem10319, 4-trifluoromethyl piperidine, hcl, 4-trifluoromethylpiperidine-hcl salt
PubChem CID 16218160
IUPAC Name 4-(trifluoromethyl)piperidine;hydrochloride
SMILES C1CNCCC1C(F)(F)F.Cl

Specifications

Melting Point ≥160°C (decomposition)
Quantity 250 mg
Beilstein 1341222
Formula Weight 189.61
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)piperidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only