missing translation for 'onlineSavingsMsg'
Learn More

[4-(Trifluoromethyl)phenyl]methanesulfonyl chloride, ≥95%, Thermo Scientific™

Chemical Identifiers

CAS 163295-75-8
Molecular Formula C8H6ClF3O2S
Molecular Weight (g/mol) 258.64
MDL Number MFCD01631932
InChI Key KKBNUPMMAGEQAT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenyl methanesulfonyl chloride, 4-trifluoromethylbenzylsulfonyl chloride, 4-trifluoromethyl benzylsulfonyl chloride, 4-trifluoromethyl phenyl methyl sulfonyl chloride, 4-chlorosulphonyl methyl benzotrifluoride, 4-trifluoromethyl-alpha-toluenesulfonyl chloride, 4-trifluoromethylphenyl methanesulfonyl chloride, 4-trifluoromethyl-phenyl-methanesulfonyl chloride, alpha-chlorosulphonyl-4-trifluoromethyl toluene
PubChem CID 594332
IUPAC Name [4-(trifluoromethyl)phenyl]methanesulfonyl chloride
SMILES FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
MO07768CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

163295-75-8
258.64
KKBNUPMMAGEQAT-UHFFFAOYSA-N
594332
FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1
C8H6ClF3O2S
MFCD01631932
4-trifluoromethyl phenyl methanesulfonyl chloride, 4-trifluoromethylbenzylsulfonyl chloride, 4-trifluoromethyl benzylsulfonyl chloride, 4-trifluoromethyl phenyl methyl sulfonyl chloride, 4-chlorosulphonyl methyl benzotrifluoride, 4-trifluoromethyl-alpha-toluenesulfonyl chloride, 4-trifluoromethylphenyl methanesulfonyl chloride, 4-trifluoromethyl-phenyl-methanesulfonyl chloride, alpha-chlorosulphonyl-4-trifluoromethyl toluene
[4-(trifluoromethyl)phenyl]methanesulfonyl chloride

Specifications

163295-75-8
95+%
MFCD01631932
KKBNUPMMAGEQAT-UHFFFAOYSA-N
[4-(trifluoromethyl)phenyl]methanesulfonyl chloride
594332
95%
95.0
C8H6ClF3O2S
4-trifluoromethyl phenyl methanesulfonyl chloride, 4-trifluoromethylbenzylsulfonyl chloride, 4-trifluoromethyl benzylsulfonyl chloride, 4-trifluoromethyl phenyl methyl sulfonyl chloride, 4-chlorosulphonyl methyl benzotrifluoride, 4-trifluoromethyl-alpha-toluenesulfonyl chloride, 4-trifluoromethylphenyl methanesulfonyl chloride, 4-trifluoromethyl-phenyl-methanesulfonyl chloride, alpha-chlorosulphonyl-4-trifluoromethyl toluene
FC(F)(F)C1=CC=C(CS(Cl)(=O)=O)C=C1
258.64
258.65
[4-(Trifluoromethyl)phenyl]methanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents