missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate 97.0+%, TCI America™
Supplier: TCI America T36221G
Specifications
| [(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate | |
| 171°C | |
| MFCD30725767 | |
| [(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium Tosylate | |
| CC1=CC=C(C=C1)S([O-])(=O)=O.COC1=CC(OC)=C([I+]C2=CC=C(C=C2)C(F)(F)F)C(OC)=C1 | |
| 610.38 | |
| 610.38 | |
| Crystalline Powder |
| 1868173-15-2 | |
| C23H22F3IO6S | |
| 1 g | |
| RTMJODUYHFSMBJ-UHFFFAOYSA-M | |
| [4-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodanium 4-methylbenzene-1-sulfonate | |
| 131697921 | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 1868173-15-2 | |
| 610.38 | |
| RTMJODUYHFSMBJ-UHFFFAOYSA-M | |
| 131697921 | |
| CC1=CC=C(C=C1)S([O-])(=O)=O.COC1=CC(OC)=C([I+]C2=CC=C(C=C2)C(F)(F)F)C(OC)=C1 |
| C23H22F3IO6S | |
| MFCD30725767 | |
| [(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium Tosylate | |
| [4-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodanium 4-methylbenzene-1-sulfonate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator