missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)imidazole, 98%

Catalog No. AAH6471106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6471106 5 g
AAH6471103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6471106 Supplier Thermo Scientific Chemicals Supplier No. H6471106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 33468-69-8 | C4H3F3N2 | 136.077 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33468-69-8
Molecular Formula C4H3F3N2
Molecular Weight (g/mol) 136.077
MDL Number MFCD08458860
InChI Key DFLGRTIPTPCKPJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1h-imidazole, 4-trifluoromethylimidazole, 4-trifluoromethyl imidazole, 5-trifluoromethyl-1h-imidazole, 4 5-trifluoromethyl imidazole, 1h-imidazole, 4-trifluoromethyl, imidazole, 4-trifluoromethyl, pubchem19080, acmc-1ailu
PubChem CID 585891
IUPAC Name 5-(trifluoromethyl)-1H-imidazole
SMILES C1=C(NC=N1)C(F)(F)F

Specifications

Melting Point 148°C to 150°C
Quantity 5 g
Formula Weight 136.08
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(Trifluoromethyl)imidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only