missing translation for 'onlineSavingsMsg'
Learn More

[[4-(Trifluoromethyl)cyclohexyl]methyl]amine (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. T28961G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28961G Supplier TCI America Supplier No. T28961G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 361393-85-3
Molecular Formula C8H14F3N
Molecular Weight (g/mol) 181.202
MDL Number MFCD07777219
InChI Key XAKIXQQQAQHCMN-UHFFFAOYSA-N
Synonym 1-Aminomethyl-4-trifluoromethylcyclohexane
PubChem CID 22337633
IUPAC Name [4-(trifluoromethyl)cyclohexyl]methanamine
SMILES C1CC(CCC1CN)C(F)(F)F

Specifications

Color Colorless
Quantity 1 g
UN Number 2735
Formula Weight 181.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material [[4-(Trifluoromethyl)cyclohexyl]methyl]amine (cis- and trans- mixture)