missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. T24175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24175G 5 g
T24171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24175G Supplier TCI America Supplier No. T24175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95233-30-0
Molecular Formula C8H11F3O2
Molecular Weight (g/mol) 196.17
MDL Number MFCD00220049,MFCD09037363,MFCD09037361
InChI Key LMEAZIIFLVDISW-UHFFFAOYSA-N
Synonym 4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexanecarboxylic acid, cis-4-trifluoromethyl cyclohexanecarboxylic acid, 4-trifluoromethyl cyclohexane-1-carboxylic acid, 1r,4r-4-trifluoromethyl cyclohexanecarboxylic acid, trans-4-trifluoromethyl cyclohexane-carboxylicacid, trans-4-trifluoromethyl cyclohexane-carboxylic acid, trans-4-trifluoromethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-cyclohexanecarboxylic acid
PubChem CID 2779182
IUPAC Name 4-(trifluoromethyl)cyclohexane-1-carboxylic acid
SMILES OC(=O)C1CCC(CC1)C(F)(F)F

Specifications

Melting Point 75°C
Color White
Quantity 5 g
Formula Weight 196.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)cyclohexanecarboxylic Acid (cis- and trans- mixture)