missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)cinnamic Acid 98.0+%, TCI America™
Supplier: TCI America T249925G
Specifications
| 4-(Trifluoromethyl)cinnamic Acid | |
| 231°C | |
| C10H6F3O2 | |
| 25 g | |
| ANRMAUMHJREENI-ZZXKWVIFSA-M | |
| (2E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoate | |
| 688070 | |
| 216.16 | |
| Crystalline Powder |
| 16642-92-5 | |
| White-Yellow | |
| MFCD00002696 | |
| 4-trifluoromethyl cinnamic acid, trans-4-trifluoromethyl cinnamic acid, p-trifluoromethyl cinnamic acid, e-3-4-trifluoromethyl phenyl acrylic acid, 2e-3-4-trifluoromethyl phenyl prop-2-enoic acid, 4-trifluoromethylcinnamic acid, 2-propenoic acid, 3-4-trifluoromethyl phenyl, 3-4-trifluoromethyl-phenyl-acrylic acid, ptf-cna, trans-4-trifluoromethylcinnamic acid | |
| [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F | |
| 215.15 | |
| CHEBI:60705 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 16642-92-5 | |
| 215.15 | |
| ANRMAUMHJREENI-ZZXKWVIFSA-M | |
| 688070 | |
| (2E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoate |
| C10H6F3O2 | |
| MFCD00002696 | |
| 4-trifluoromethyl cinnamic acid, trans-4-trifluoromethyl cinnamic acid, p-trifluoromethyl cinnamic acid, e-3-4-trifluoromethyl phenyl acrylic acid, 2e-3-4-trifluoromethyl phenyl prop-2-enoic acid, 4-trifluoromethylcinnamic acid, 2-propenoic acid, 3-4-trifluoromethyl phenyl, 3-4-trifluoromethyl-phenyl-acrylic acid, ptf-cna, trans-4-trifluoromethylcinnamic acid | |
| CHEBI:60705 | |
| [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F |
Safety and Handling
TSCA : No