missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)cinnamic Acid 98.0+%, TCI America™

Catalog No. T24995G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24995G 5 g
T24991G 1 g
T249925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T24995G Supplier TCI America Supplier No. T24995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16642-92-5
Molecular Formula C10H6F3O2
Molecular Weight (g/mol) 215.15
MDL Number MFCD00002696
InChI Key ANRMAUMHJREENI-ZZXKWVIFSA-M
Synonym 4-trifluoromethyl cinnamic acid, trans-4-trifluoromethyl cinnamic acid, p-trifluoromethyl cinnamic acid, e-3-4-trifluoromethyl phenyl acrylic acid, 2e-3-4-trifluoromethyl phenyl prop-2-enoic acid, 4-trifluoromethylcinnamic acid, 2-propenoic acid, 3-4-trifluoromethyl phenyl, 3-4-trifluoromethyl-phenyl-acrylic acid, ptf-cna, trans-4-trifluoromethylcinnamic acid
PubChem CID 688070
ChEBI CHEBI:60705
IUPAC Name (2E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoate
SMILES [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 231°C
Color White-Yellow
Quantity 5 g
Formula Weight 216.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)cinnamic Acid