missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Catalog No. AC188460010
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC188460010 1g Glass bottle
AC188460050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC188460010 Supplier Thermo Scientific Chemicals Supplier No. 188460010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 455-24-3
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00002562
InChI Key SWKPKONEIZGROQ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoic acid, p-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-p-toluic acid, benzoic acid, 4-trifluoromethyl, p-carboxybenzotrifluoride, 4-trifluoromethyl-benzoic acid, a,a,a-trifluoro-p-toluic acid, ptf-boa, 4-carboxybenzotrifluoride
PubChem CID 9966
ChEBI CHEBI:60696
IUPAC Name 4-(trifluoromethyl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CF3C6H4CO2H
Quantity 1g
Beilstein 09, III, 2355
Formula Weight 190.12
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-242-8
TSCA TSCA

RUO – Research Use Only