missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethyl)acetophenone 98.0+%, TCI America™

Catalog No. T17841G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17841G 1 g
T17845G 5 g
T178425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T17841G Supplier TCI America Supplier No. T17841G

Chemical Identifiers

CAS 709-63-7
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000401
InChI Key HHAISVSEJFEWBZ-UHFFFAOYSA-N
Synonym 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl
PubChem CID 69731
IUPAC Name 1-[4-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Color White-Yellow
Boiling Point 80°C
Quantity 1 g
Formula Weight 188.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-(Trifluoromethyl)acetophenone