missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethyl)acetophenone, 98+%

Catalog No. AAA1552906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1552906 5 g
AAA1552914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1552906 Supplier Thermo Scientific Chemicals Supplier No. A1552906

CAS: 709-63-7 | C9H7F3O | 188.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 709-63-7
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000401
InChI Key HHAISVSEJFEWBZ-UHFFFAOYSA-N
Synonym 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl
PubChem CID 69731
IUPAC Name 1-[4-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 29°C to 35°C
Density 0.924
Boiling Point 83°C to 85°C (9 mmHg)
Flash Point 84°C (183°F)
Odor Characteristic
Refractive Index 1.462
Quantity 5 g
Beilstein 1870425
Formula Weight 188.15
Percent Purity ≥98%
Chemical Name or Material 4'-(Trifluoromethyl)acetophenone
Show More Show Less
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 211-913-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only