missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)-1H-pyrazole, 97%

Catalog No. AC446490010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446490010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC446490010 Supplier Thermo Scientific Chemicals Supplier No. 446490010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52222-73-8 | C4H3F3N2 | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52222-73-8
Molecular Formula C4H3F3N2
Molecular Weight (g/mol) 136.08
InChI Key KDEJQUNODYXYBJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1h-pyrazole, 1h-pyrazole, 4-trifluoromethyl, 4-trifluoromethylpyrazole, 4-trifluoromethyl pyrazole, 4-trifluoromethyl-1 h-pyrazole, 4-trifluoromethyl ;-1h-pyrazole
PubChem CID 12777795
IUPAC Name 4-(trifluoromethyl)-1H-pyrazole
SMILES C1=C(C=NN1)C(F)(F)F

Specifications

Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 136.08
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)-1H-pyrazole
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only