missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl Isocyanate 98.0+%, TCI America™

Catalog No. T24875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24875G 5 g
T248725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24875G Supplier TCI America Supplier No. T24875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35037-73-1
Molecular Formula C8H4F3NO2
Molecular Weight (g/mol) 203.12
MDL Number MFCD00035702
InChI Key LGPKFIGMLPDYEA-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
PubChem CID 92298
IUPAC Name 1-isocyanato-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1N=C=O)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
UN Number 2206
Formula Weight 203.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)phenyl Isocyanate