missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)phenyl isocyanate, 97%
Description
4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine
Solubility
Reacts with water.
Notes
Moisture Sensitive. Protect from heat. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas. Incompatible with alcohols, amines and heat.
4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine
Solubility
Reacts with water.
Notes
Moisture Sensitive. Protect from heat. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas. Incompatible with alcohols, amines and heat.
Specifications
Specifications
| Melting Point | -29°C |
| Density | 1.346 |
| Boiling Point | 78°C (22 mmHg) |
| Flash Point | 70°C (158°F) |
| Refractive Index | 1.458 |
| Quantity | 1 g |
| UN Number | UN2206 |
| Beilstein | 977669 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| Show More |
Safety and Handling
| Hazard Category | H227-H302-H311-H314-H317-H330-H334-H335 |
| Hazard Statement | GHS H Statement H331-H334-H227-H302-H315-H319-H335 Toxic if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Combustible liquid. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: I |
| EINECSNumber | 252-328-0 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only