missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl isocyanate, 97%

Catalog No. AAB2003314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2003314 25 g
AAB2003303 1 g
AAB2003306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2003314 Supplier Thermo Scientific Chemicals Supplier No. B2003314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35037-73-1 | C8H4F3NO2 | 203.12 g/mol

4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine

Solubility
Reacts with water.

Notes
Moisture Sensitive. Protect from heat. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas. Incompatible with alcohols, amines and heat.

Chemical Identifiers

CAS 35037-73-1
Molecular Formula C8H4F3NO2
Molecular Weight (g/mol) 203.12
MDL Number MFCD00035702
InChI Key LGPKFIGMLPDYEA-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
PubChem CID 92298
IUPAC Name 1-isocyanato-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1N=C=O)OC(F)(F)F

Specifications

Melting Point -29°C
Density 1.346
Boiling Point 78°C (22 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.458
Quantity 25 g
UN Number UN2206
Beilstein 977669
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 203.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)phenyl isocyanate
Show More Show Less
Hazard Category H227-H302-H311-H314-H317-H330-H334-H335
Hazard Statement GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 252-328-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only