missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl isocyanate, 97%, Thermo Scientific™

Catalog No. p-2699874
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
Catalog No. Quantity
SB01053EA 10 g
1 options

Catalog No. SB01053EA

Supplier: Thermo Scientific Chemicals SB01053EA

Only null left

Chemical Identifiers

CAS 35037-73-1
Molecular Formula C8H4F3NO2
Molecular Weight (g/mol) 203.12
MDL Number MFCD00035702
InChI Key LGPKFIGMLPDYEA-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
PubChem CID 92298
IUPAC Name 1-isocyanato-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1N=C=O)OC(F)(F)F

Specifications

CAS Min % 97.0
Density 1.34g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4NCO
Quantity 10 g
Formula Weight 203.12
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)phenyl isocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.