missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phényl isocyanate, 97%, Thermo Scientific™

Numéro de catalogue. SB01053EA
Modifier l'affichage
Click to view available options
Quantité:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
SB01053EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. SB01053EA Fournisseur Thermo Scientific Chemicals Code fournisseur SB01053EA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 35037-73-1
Formule moléculaire C8H4F3NO2
Poids moléculaire (g/mol) 203.12
Numéro MDL MFCD00035702
Clé InChI LGPKFIGMLPDYEA-UHFFFAOYSA-N
Synonyme 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
PubChem CID 92298
Nom de l’IUPAC 1-isocyanato-4-(trifluoromethoxy)benzène
SOURIRES C1=CC(=CC=C1N=C=O)OC(F)(F)F

Spécifications

Nom ou substance chimique 4-(Trifluoromethoxy)phenyl isocyanate
CAS % minimum 97.0
Densité 1.34g/mL
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Formule linéaire CF3OC6H4NCO
Quantité 10 g
Poids de formule 203.12
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.