missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Trifluoromethoxy)benzoyl Chloride 98.0+%, TCI America™
$46.44 - $144.33
Chemical Identifiers
CAS | 36823-88-8 |
---|---|
Molecular Formula | C8H4ClF3O2 |
Molecular Weight (g/mol) | 224.563 |
MDL Number | MFCD00052329 |
InChI Key | ZXKKOFJYPRJFIE-UHFFFAOYSA-N |
Synonym | 4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride |
PubChem CID | 142110 |
IUPAC Name | 4-(trifluoromethoxy)benzoyl chloride |
SMILES | C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F |
Chemical Identifiers
36823-88-8 | |
224.563 | |
ZXKKOFJYPRJFIE-UHFFFAOYSA-N | |
142110 | |
C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F |
C8H4ClF3O2 | |
MFCD00052329 | |
4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride | |
4-(trifluoromethoxy)benzoyl chloride |
Specifications
36823-88-8 | |
56°C | |
MFCD00052329 | |
3265 | |
ZXKKOFJYPRJFIE-UHFFFAOYSA-N | |
4-(trifluoromethoxy)benzoyl chloride | |
142110 | |
≥98.0% (GC,T) | |
4-(Trifluoromethoxy)benzoyl Chloride |
Colorless | |
C8H4ClF3O2 | |
1 g | |
4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride | |
C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F | |
224.563 | |
224.56 | |
Liquid |
Safety and Handling
TSCA : No