missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzoyl chloride, 98%

CAS: 36823-88-8 | C8H4ClF3O2 | 224.56 g/mol

$68.63 - $68.63

Chemical Identifiers

CAS 36823-88-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD00052329
InChI Key ZXKKOFJYPRJFIE-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride
PubChem CID 142110
IUPAC Name 4-(trifluoromethoxy)benzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC306350010
SDS View Documents
Thermo Scientific Chemicals
306350010
1 g Glass bottle
Each for $68.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

36823-88-8
224.56
ZXKKOFJYPRJFIE-UHFFFAOYSA-N
142110
C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F
C8H4ClF3O2
MFCD00052329
4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride
4-(trifluoromethoxy)benzoyl chloride

Specifications

36823-88-8
100.0
1.4200g/mL
32°C
97.5% min. (GC)
C8H4ClF3O2
CF3OC6H4COCl
1 g
4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride
C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F
224.56
224.56
Liquid
97.5
Colorless to Pink
55°C to 56°C (2.0 mmHg)
Authentic
Glass bottle
1.4710 to 1.473
MFCD00052329
1.42
ZXKKOFJYPRJFIE-UHFFFAOYSA-N
4-(trifluoromethoxy)benzoyl chloride
142110
98%
4-(Trifluoromethoxy)benzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only